accessibility menu, dialog, popup

UserName

ONOONClO

CAS RN 54455-34-4

IUPAC Name: 2-(chloromethyl)-4-nitro-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(chloromethyl)-4-nitroisoindole-1,3-dione
Molecular Weight (g/mol): 240.6
Molecular Formula: C9H5ClN2O4
InChi Key: WECOPMBZMWMHAB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC2=C1C(=O)N(CCl)C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results