missing translation for 'onlineSavingsMsg'
Learn More

N-Chloroacetyl-DL-phenylalanine 97.0+%, TCI America™

Catalog No. C01031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C01031G Supplier TCI America Supplier No. C01031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7765-11-9
Molecular Formula C11H12ClNO3
Molecular Weight (g/mol) 241.67
MDL Number MFCD00037263
InChI Key FUHGSOAURCZWCC-UHFFFAOYNA-N
PubChem CID 98109
IUPAC Name 2-[(2-chloroacetyl)amino]-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC(=O)CCl

Specifications

Melting Point 127°C
Quantity 1 g
Formula Weight 241.67
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetyl-DL-phenylalanine