accessibility menu, dialog, popup

UserName

CAS RN 7765-11-9

IUPAC Name: 2-(2-chloroacetamido)-3-phenylpropanoic acid
Synonyms: 2-(2-chloroacetamido)-3-phenylpropanoic acid
Molecular Weight (g/mol): 241.67
Molecular Formula: C11H12ClNO3
InChi Key: FUHGSOAURCZWCC-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NC(=O)CCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results