missing translation for 'onlineSavingsMsg'
Learn More

N-Chloroacetyl-DL-alanine 98.0+%, TCI America™

Catalog No. C00971G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C00971G Supplier TCI America Supplier No. C00971G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1190-32-5
Molecular Formula C5H8ClNO3
Molecular Weight (g/mol) 165.573
MDL Number MFCD00020401
InChI Key HTAQFYLADZNZHZ-UHFFFAOYSA-N
PubChem CID 102573
IUPAC Name 2-[(2-chloroacetyl)amino]propanoic acid
SMILES CC(C(=O)O)NC(=O)CCl

Specifications

Melting Point 125°C
Color White
Quantity 1 g
Formula Weight 165.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetyl-DL-alanine