accessibility menu, dialog, popup

UserName

CH3NHOClOHO

CAS RN 1190-32-5

IUPAC Name: 2-(2-chloroacetamido)propanoic acid
Synonyms: 2-(2-chloroacetamido)propanoic acid
Molecular Weight (g/mol): 165.57
Molecular Formula: C5H8ClNO3
InChi Key: HTAQFYLADZNZHZ-UHFFFAOYNA-N
SMILES: CC(NC(=O)CCl)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results