missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ N-Carbobenzyloxy-L-serine, 99+%

Chemical Identifiers

CAS 1145-80-8
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.23
MDL Number MFCD00002662
InChI Key GNIDSOFZAKMQAO-UHFFFAOYNA-N
Synonym z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
PubChem CID 100310
IUPAC Name (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229510050
SDS View Documents
Thermo Scientific™
229510050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC229510250
SDS View Documents
Thermo Scientific™
229510250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1145-80-8
239.23
GNIDSOFZAKMQAO-UHFFFAOYNA-N
100310
OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
C11H13NO5
MFCD00002662
z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
(2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

1145-80-8
100.0
99% min. (HPLC)
C11H13NO5
MFCD00002662
06, IV, 2374
z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
GNIDSOFZAKMQAO-UHFFFAOYNA-N
(2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
100310
99+%
99.0
Authentic
Glass bottle
HOCH2CH(NHCO2CH2C6H5)CO2H
5g
+ 6
Solubility in water: .. Other solubilities: soluble in most common organic solvents
OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
239.23
239.23
N-Carbobenzyloxy-L-serine, 99+%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-546-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only