missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Carbobenzyloxy-L-serine, 99+%

Catalog No. AC229510050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229510050 5 g Glass bottle
AC229510250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC229510050 Supplier Thermo Scientific Chemicals Supplier No. 229510050

CAS: 1145-80-8 | C11H13NO5 | 239.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1145-80-8
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.23
MDL Number MFCD00002662
InChI Key GNIDSOFZAKMQAO-UHFFFAOYNA-N
Synonym z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
PubChem CID 100310
IUPAC Name (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 115°C to 120°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula HOCH2CH(NHCO2CH2C6H5)CO2H
Quantity 5 g
Beilstein 06, IV, 2374
Specific Rotation + 6
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 239.23
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material N-Carbobenzyloxy-L-serine, 99+%
Show More Show Less
EINECSNumber 214-546-4

RUO – Research Use Only