missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxyoxysuccinimide 98.0+%, TCI America™

Catalog No. p-7124804
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C112425G 25 g
C1124250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C112425G Supplier TCI America Supplier No. C112425G
Only null left

Chemical Identifiers

CAS 13139-17-8
Molecular Formula C12H11NO5
Molecular Weight (g/mol) 249.222
MDL Number MFCD00005513
InChI Key MJSHDCCLFGOEIK-UHFFFAOYSA-N
Synonym n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide
PubChem CID 83172
IUPAC Name benzyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 81°C
Color White
Quantity 25 g
Formula Weight 249.22
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxyoxysuccinimide