accessibility menu, dialog, popup

UserName

OOONOO

CAS RN 13139-17-8

IUPAC Name: benzyl 2,5-dioxopyrrolidin-1-yl carbonate
Synonyms: benzyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular Weight (g/mol): 249.22
Molecular Formula: C12H11NO5
InChi Key: MJSHDCCLFGOEIK-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)