missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxyglycine Methyl Ester 95.0+%, TCI America™

Catalog No. C28385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28385G 5 g
C283825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28385G Supplier TCI America Supplier No. C28385G

Chemical Identifiers

CAS 1212-53-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00043631
InChI Key DZYBBBYFLOPVOL-UHFFFAOYSA-N
Synonym z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester
PubChem CID 71033
IUPAC Name methyl 2-{[(benzyloxy)carbonyl]amino}acetate
SMILES COC(=O)CNC(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 151°C
Quantity 5 g
Formula Weight 223.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Carbobenzoxyglycine Methyl Ester