accessibility menu, dialog, popup

UserName

H3COONHOO

CAS RN 1212-53-9

IUPAC Name: methyl 2-{[(benzyloxy)carbonyl]amino}acetate
Synonyms: methyl 2-{[(benzyloxy)carbonyl]amino}acetate
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: DZYBBBYFLOPVOL-UHFFFAOYSA-N
SMILES: COC(=O)CNC(=O)OCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)