missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-O-tert-butyl-L-serine 98.0+%, TCI America™
$32.28 - $104.43
Chemical Identifiers
| CAS | 1676-75-1 |
|---|---|
| Molecular Formula | C15H21NO5 |
| Molecular Weight (g/mol) | 295.335 |
| MDL Number | MFCD00038275 |
| InChI Key | TXDGEONUWGOCJG-LBPRGKRZSA-N |
| Synonym | z-ser tbu-oh, n-cbz-o-tert-butyl-l-serine, z-ser-otbu, n-carbobenzoxy-o-tert-butyl-l-serine, s-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-alpha-benzyloxycarbonyl-o-t-butyl-l-serine, s-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, 2s-3-2-methylpropan-2-yl oxy-2-phenylmethoxycarbonylamino propanoic acid, 2s-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, z-ser t-bu-oh |
| PubChem CID | 1715617 |
| IUPAC Name | (2S)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid |
| SMILES | CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| C15H21NO5 | |
| MFCD00038275 | |
| z-ser tbu-oh, n-cbz-o-tert-butyl-l-serine, z-ser-otbu, n-carbobenzoxy-o-tert-butyl-l-serine, s-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-alpha-benzyloxycarbonyl-o-t-butyl-l-serine, s-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, 2s-3-2-methylpropan-2-yl oxy-2-phenylmethoxycarbonylamino propanoic acid, 2s-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, z-ser t-bu-oh | |
| (2S)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid |
Specifications
| 86°C | |
| White | |
| MFCD00038275 | |
| TXDGEONUWGOCJG-LBPRGKRZSA-N | |
| (2S)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid | |
| 1715617 | |
| ≥98.0% (T) | |
| N-Carbobenzoxy-O-tert-butyl-L-serine |
| 1676-75-1 | |
| C15H21NO5 | |
| z-ser tbu-oh, n-cbz-o-tert-butyl-l-serine, z-ser-otbu, n-carbobenzoxy-o-tert-butyl-l-serine, s-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-alpha-benzyloxycarbonyl-o-t-butyl-l-serine, s-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, 2s-3-2-methylpropan-2-yl oxy-2-phenylmethoxycarbonylamino propanoic acid, 2s-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, z-ser t-bu-oh | |
| CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
| 295.335 | |
| 295.34 | |
| Crystalline Powder |
Safety and Handling
TSCA : No