missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-O-tert-butyl-L-serine 98.0+%, TCI America™

Catalog No. C13811G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13811G 1 g
C13815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13811G Supplier TCI America Supplier No. C13811G
Only null left

Chemical Identifiers

CAS 1676-75-1
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00038275
InChI Key TXDGEONUWGOCJG-LBPRGKRZSA-N
Synonym z-ser tbu-oh, n-cbz-o-tert-butyl-l-serine, z-ser-otbu, n-carbobenzoxy-o-tert-butyl-l-serine, s-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-alpha-benzyloxycarbonyl-o-t-butyl-l-serine, s-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, 2s-3-2-methylpropan-2-yl oxy-2-phenylmethoxycarbonylamino propanoic acid, 2s-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, z-ser t-bu-oh
PubChem CID 1715617
IUPAC Name (2S)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 86°C
Color White
Quantity 1 g
Formula Weight 295.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-O-tert-butyl-L-serine