missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-N-methyl-L-phenylalanine 98.0+%, TCI America™
$105.69 - $342.69
Chemical Identifiers
| CAS | 2899-07-2 |
|---|---|
| Molecular Formula | C18H19NO4 |
| Molecular Weight (g/mol) | 313.35 |
| MDL Number | MFCD00153393 |
| InChI Key | JDGXKNMKNMSHRJ-UHFFFAOYNA-N |
| Synonym | N-Cbz-N-methyl-L-phenylalanine, Z-N-Me-Phe-OH |
| PubChem CID | 6544472 |
| IUPAC Name | (2S)-2-[methyl(phenylmethoxycarbonyl)amino]-3-phenylpropanoic acid |
| SMILES | CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2=CC=CC=C2 |
Chemical Identifiers
| 2899-07-2 | |
| 313.35 | |
| JDGXKNMKNMSHRJ-UHFFFAOYNA-N | |
| 6544472 | |
| CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2=CC=CC=C2 |
| C18H19NO4 | |
| MFCD00153393 | |
| N-Cbz-N-methyl-L-phenylalanine, Z-N-Me-Phe-OH | |
| (2S)-2-[methyl(phenylmethoxycarbonyl)amino]-3-phenylpropanoic acid |
Specifications
| 2899-07-2 | |
| White-Yellow | |
| MFCD00153393 | |
| N-Cbz-N-methyl-L-phenylalanine, Z-N-Me-Phe-OH | |
| CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2=CC=CC=C2 | |
| 313.35 | |
| 313.35 | |
| Crystalline Powder |
| 69°C | |
| C18H19NO4 | |
| 1 g | |
| JDGXKNMKNMSHRJ-UHFFFAOYNA-N | |
| (2S)-2-[methyl(phenylmethoxycarbonyl)amino]-3-phenylpropanoic acid | |
| 6544472 | |
| ≥98.0% (HPLC,T) | |
| N-Carbobenzoxy-N-methyl-L-phenylalanine |
Safety and Handling
TSCA : No