CAS RN 2899-07-2
CAS RN 2899-07-2
IUPAC Name:
2-{[(benzyloxy)carbonyl](methyl)amino}-3-phenylpropanoic acid
Synonyms:
2-{[(benzyloxy)carbonyl](methyl)amino}-3-phenylpropanoic acid
Molecular Weight (g/mol):
313.35
Molecular Formula:
C18H19NO4
InChi Key:
JDGXKNMKNMSHRJ-UHFFFAOYNA-N
SMILES:
CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N-Carbobenzoxy-N-methyl-L-phenylalanine 98.0+%, TCI America™
CAS: 2899-07-2 Molecular Formula: C18H19NO4 Molecular Weight (g/mol): 313.35 MDL Number: MFCD00153393 InChI Key: JDGXKNMKNMSHRJ-UHFFFAOYNA-N Synonym: N-Cbz-N-methyl-L-phenylalanine, Z-N-Me-Phe-OH PubChem CID: 6544472 IUPAC Name: (2S)-2-[methyl(phenylmethoxycarbonyl)amino]-3-phenylpropanoic acid SMILES: CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2=CC=CC=C2