missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-N-methyl-L-phenylalanine 98.0+%, TCI America™

Catalog No. C24681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24681G 1 g
C24685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24681G Supplier TCI America Supplier No. C24681G
Only null left

Chemical Identifiers

CAS 2899-07-2
Molecular Formula C18H19NO4
Molecular Weight (g/mol) 313.35
MDL Number MFCD00153393
InChI Key JDGXKNMKNMSHRJ-UHFFFAOYNA-N
Synonym N-Cbz-N-methyl-L-phenylalanine, Z-N-Me-Phe-OH
PubChem CID 6544472
IUPAC Name (2S)-2-[methyl(phenylmethoxycarbonyl)amino]-3-phenylpropanoic acid
SMILES CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 1 g
Formula Weight 313.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-N-methyl-L-phenylalanine