missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-valine 99.0+%, TCI America™

Catalog No. p-7124593
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C07615G 5 g
C076125G 25 g
2 options

Catalog No. C07615G

Supplier: TCI America C07615G

Only null left

Chemical Identifiers

CAS 1149-26-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00008922
InChI Key CANZBRDGRHNSGZ-NSHDSACASA-N
Synonym n-carbobenzyloxy-l-valine, n-cbz-l-valine, z-val-oh, cbz-l-valine, n-benzyloxycarbonyl-l-valine, n-carbobenzoxy-l-valine, z-l-val-oh, carbobenzoxy-l-valine, z-valine, carbobenzyloxy-l-valine
PubChem CID 726987
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 60°C
Color White
Quantity 5 g
Formula Weight 251.28
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-valine
EINECSNumber (3)-4273
TSCA No