accessibility menu, dialog, popup

UserName

H3C(S)OHHNCH3OOO

CAS RN 1149-26-4

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid|Z-VAL-OH
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: CANZBRDGRHNSGZ-NSHDSACASA-N
SMILES: CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)