missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-L-tyrosine 98.0+%, TCI America™
Supplier: TCI America C17295G
Specifications
| N-Carbobenzoxy-L-tyrosine | |
| 95°C | |
| C17H17NO5 | |
| 5 g | |
| MCRMUCXATQAAMN-GGYSOQFKNA-N | |
| (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid | |
| 712438 | |
| ≥98.0% (T) |
| 1164-16-5 | |
| White-Yellow | |
| MFCD00037180 | |
| z-tyr-oh, n-cbz-l-tyrosine, n-benzyloxycarbonyl-l-tyrosine, cbz-l-tyrosine, n-carbobenzoxy-l-tyrosine, n-carbobenzyloxy-l-tyrosine, cbz-tyr-oh, benzyloxycarbonyl-l-tyrosine, n-cbz-l-tyrosine-oh, z-tyrosine | |
| OC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1 | |
| 315.33 | |
| 315.33 | |
| Crystalline Powder |
Chemical Identifiers
| 1164-16-5 | |
| 315.33 | |
| MCRMUCXATQAAMN-GGYSOQFKNA-N | |
| 712438 | |
| OC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1 |
| C17H17NO5 | |
| MFCD00037180 | |
| z-tyr-oh, n-cbz-l-tyrosine, n-benzyloxycarbonyl-l-tyrosine, cbz-l-tyrosine, n-carbobenzoxy-l-tyrosine, n-carbobenzyloxy-l-tyrosine, cbz-tyr-oh, benzyloxycarbonyl-l-tyrosine, n-cbz-l-tyrosine-oh, z-tyrosine | |
| (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid |
Safety and Handling
TSCA : No