missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-tyrosine 98.0+%, TCI America™

Catalog No. p-7125140
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17295G 5 g
C172925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17295G Supplier TCI America Supplier No. C17295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1164-16-5
Molecular Formula C17H17NO5
Molecular Weight (g/mol) 315.33
MDL Number MFCD00037180
InChI Key MCRMUCXATQAAMN-GGYSOQFKNA-N
Synonym z-tyr-oh, n-cbz-l-tyrosine, n-benzyloxycarbonyl-l-tyrosine, cbz-l-tyrosine, n-carbobenzoxy-l-tyrosine, n-carbobenzyloxy-l-tyrosine, cbz-tyr-oh, benzyloxycarbonyl-l-tyrosine, n-cbz-l-tyrosine-oh, z-tyrosine
PubChem CID 712438
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 5 g
Formula Weight 315.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-tyrosine