missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-serine Methyl Ester 98.0+%, TCI America™

Catalog No. C14035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14035G 5 g
C140325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14035G Supplier TCI America Supplier No. C14035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1676-81-9
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00077039
InChI Key CINAUOAOVQPWIB-UHFFFAOYNA-N
Synonym z-ser-ome, n-cbz-l-serine methyl ester, cbz-ser-ome, n-carbobenzoxy-l-serine methyl ester, z-l-serine methyl ester, cbz-l-serine methyl ester, n-z-l-serine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate, n-carbobenzyloxy-l-serine methyl ester, d-n-cbz-serine methyl ester
PubChem CID 6999535
IUPAC Name methyl 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoate
SMILES COC(=O)C(CO)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 37°C
Color White-Yellow
Quantity 5 g
Formula Weight 253.25
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-serine Methyl Ester
TSCA No
Recommended Storage Refrigerator