accessibility menu, dialog, popup

UserName

H3COOOHNHOO

CAS RN 1676-81-9

IUPAC Name: methyl 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoate
Synonyms: methyl 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoate
Molecular Weight (g/mol): 253.25
Molecular Formula: C12H15NO5
InChi Key: CINAUOAOVQPWIB-UHFFFAOYNA-N
SMILES: COC(=O)C(CO)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results