missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-serine Benzyl Ester 97.0+%, TCI America™

Catalog No. C28735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28735G 5 g
C287325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28735G Supplier TCI America Supplier No. C28735G

Chemical Identifiers

CAS 21209-51-8
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.352
MDL Number MFCD00037824
InChI Key MHHDPGHZHFJLBZ-INIZCTEOSA-N
Synonym z-ser-obzl, cbz-ser-obzl, n-cbz-l-serine benzyl ester, n-z-l-serine benzyl ester, s-benzyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate, n-carbobenzoxy-l-serine benzyl ester, benzyl n-benzyloxy carbonyl-l-serinate, l-serine,n-phenylmethoxy carbonyl-, phenylmethyl ester, serine, n-phenylmethoxy carbonyl-, phenylmethyl ester, z-l-serine benzyl ester
PubChem CID 6992970
IUPAC Name benzyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
SMILES C1=CC=C(C=C1)COC(=O)C(CO)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 85°C
Color White
Quantity 5 g
Formula Weight 329.35
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-serine Benzyl Ester