accessibility menu, dialog, popup

UserName

OOOHNHOO

CAS RN 21209-51-8

IUPAC Name: benzyl 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoate
Synonyms: benzyl 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoate
Molecular Weight (g/mol): 329.35
Molecular Formula: C18H19NO5
InChi Key: MHHDPGHZHFJLBZ-UHFFFAOYNA-N
SMILES: OCC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)