missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-L-proline 98.0+%, TCI America™
Supplier: TCI America C07135G
Specifications
| N-Carbobenzoxy-L-proline | |
| 77°C | |
| C13H15NO4 | |
| 5 g | |
| JXGVXCZADZNAMJ-NSHDSACASA-N | |
| (2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid | |
| 101987 | |
| ≥98.0% (HPLC,T) |
| 1148-11-4 | |
| White-Yellow | |
| MFCD00003170 | |
| z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline | |
| C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O | |
| 249.266 | |
| 249.27 | |
| Crystalline Powder |
Chemical Identifiers
| 1148-11-4 | |
| 249.266 | |
| JXGVXCZADZNAMJ-NSHDSACASA-N | |
| 101987 | |
| C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
| C13H15NO4 | |
| MFCD00003170 | |
| z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline | |
| (2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid |
Safety and Handling
RTECSNumber : UY0745000
TSCA : No