missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-proline 98.0+%, TCI America™

Catalog No. p-7124575
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07135G 5 g
C071325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07135G Supplier TCI America Supplier No. C07135G
Only null left

Chemical Identifiers

CAS 1148-11-4
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD00003170
InChI Key JXGVXCZADZNAMJ-NSHDSACASA-N
Synonym z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline
PubChem CID 101987
IUPAC Name (2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 5 g
Formula Weight 249.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-proline
RTECSNumber UY0745000
TSCA No