missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-L-phenylalaninol 98.0+%, TCI America™
$55.92 - $154.35
Chemical Identifiers
| CAS | 6372-14-1 |
|---|---|
| Molecular Formula | C17H19NO3 |
| Molecular Weight (g/mol) | 285.343 |
| MDL Number | MFCD00191138 |
| InChI Key | WPOFMMJJCPZPAO-INIZCTEOSA-N |
| Synonym | z-l-phenylalaninol, z-phenylalaninol, cbz-phenylalaninol, s-cbz-phenylalaninol, cbz-l-phenylalaninol, s---2-benzyloxycarbonylamino-3-phenyl-1-propanol, n-carbobenzoxy-l-phenylalaninol, s-2-cbz-amino-3-phenyl-1-propanol, s---2-carbobenzyloxyamino-3-phenyl-1-propanol, s-2-z-amino-3-phenyl-1-propanol |
| PubChem CID | 853481 |
| IUPAC Name | benzyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate |
| SMILES | C1=CC=C(C=C1)CC(CO)NC(=O)OCC2=CC=CC=C2 |
Chemical Identifiers
| 6372-14-1 | |
| 285.343 | |
| WPOFMMJJCPZPAO-INIZCTEOSA-N | |
| 853481 | |
| C1=CC=C(C=C1)CC(CO)NC(=O)OCC2=CC=CC=C2 |
| C17H19NO3 | |
| MFCD00191138 | |
| z-l-phenylalaninol, z-phenylalaninol, cbz-phenylalaninol, s-cbz-phenylalaninol, cbz-l-phenylalaninol, s---2-benzyloxycarbonylamino-3-phenyl-1-propanol, n-carbobenzoxy-l-phenylalaninol, s-2-cbz-amino-3-phenyl-1-propanol, s---2-carbobenzyloxyamino-3-phenyl-1-propanol, s-2-z-amino-3-phenyl-1-propanol | |
| benzyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate |
Specifications
| 93°C | |
| White | |
| MFCD00191138 | |
| WPOFMMJJCPZPAO-INIZCTEOSA-N | |
| benzyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate | |
| 285.343 | |
| 285.34 | |
| Crystalline Powder |
| 6372-14-1 | |
| C17H19NO3 | |
| z-l-phenylalaninol, z-phenylalaninol, cbz-phenylalaninol, s-cbz-phenylalaninol, cbz-l-phenylalaninol, s---2-benzyloxycarbonylamino-3-phenyl-1-propanol, n-carbobenzoxy-l-phenylalaninol, s-2-cbz-amino-3-phenyl-1-propanol, s---2-carbobenzyloxyamino-3-phenyl-1-propanol, s-2-z-amino-3-phenyl-1-propanol | |
| C1=CC=C(C=C1)CC(CO)NC(=O)OCC2=CC=CC=C2 | |
| 1 g | |
| 853481 | |
| ≥98.0% (HPLC) | |
| N-Carbobenzoxy-L-phenylalaninol |
Safety and Handling
TSCA : No