accessibility menu, dialog, popup

UserName

CAS RN 6372-14-1

OHNHOO

CAS RN 6372-14-1

IUPAC Name: benzyl N-(1-hydroxy-3-phenylpropan-2-yl)carbamate
Synonyms: benzyl N-(1-hydroxy-3-phenylpropan-2-yl)carbamate
Molecular Weight (g/mol): 285.34
Molecular Formula: C17H19NO3
InChi Key: WPOFMMJJCPZPAO-UHFFFAOYNA-N
SMILES: OCC(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)