missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-L-phenylalaninol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 93°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 285.34 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Carbobenzoxy-L-phenylalaninol |
Safety and Handling
| TSCA | No |