missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-methionine 98.0+%, TCI America™

Catalog No. p-7124584
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07375G 5 g
C073725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07375G Supplier TCI America Supplier No. C07375G

Chemical Identifiers

CAS 1152-62-1
Molecular Formula C13H17NO4S
Molecular Weight (g/mol) 283.34
MDL Number MFCD00065124
InChI Key FPKHNNQXKZMOJJ-UHFFFAOYNA-N
Synonym z-met-oh, n-cbz-l-methionine, cbz-l-methionine, cbz-met-oh, z-l-methionine, n-carbobenzyloxy-l-methionine, n-carbobenzoxy-l-methionine, benzyloxycarbonyl-l-methionine, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine
PubChem CID 1550857
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
SMILES CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
Formula Weight 283.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-methionine