missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-isoleucine 98.0+%, TCI America™

Catalog No. C135425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C135425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C135425G Supplier TCI America Supplier No. C135425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3160-59-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00027064
InChI Key JSHXJPFZKBRLFU-JQWIXIFHSA-N
Synonym cbz-l-isoleucine, n-cbz-l-isoleucine, z-l-isoleucine, z-ile-oh, n-carbobenzyloxy-l-isoleucine, n-carbobenzoxy-l-isoleucine, cbz-l-ile-oh, n-benzyloxycarbonyl-l-isoleucine, l-isoleucine, n-phenylmethoxy carbonyl, 2s,3s-2-benzyloxy carbonyl amino-3-methylpentanoic acid
PubChem CID 2724772
IUPAC Name (2S,3S)-3-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CCC(C)C(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 25 g
Formula Weight 265.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystals
Chemical Name or Material N-Carbobenzoxy-L-isoleucine