CAS RN 3160-59-6
IUPAC Name:
(2S,3S)-2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoate
Synonyms:
(2S,3S)-2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoate
Molecular Weight (g/mol):
264.3
Molecular Formula:
C14H18NO4
InChi Key:
JSHXJPFZKBRLFU-JQWIXIFHSA-M
SMILES:
CC[C@H](C)[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
1
–
1
of
1
results
1
N-Carbobenzoxy-L-isoleucine 98.0+%, TCI America™
CAS: 3160-59-6 Molecular Formula: C14H19NO4 Molecular Weight (g/mol): 265.309 MDL Number: MFCD00027064 InChI Key: JSHXJPFZKBRLFU-JQWIXIFHSA-N Synonym: cbz-l-isoleucine,n-cbz-l-isoleucine,z-l-isoleucine,z-ile-oh,n-carbobenzyloxy-l-isoleucine,n-carbobenzoxy-l-isoleucine,cbz-l-ile-oh,n-benzyloxycarbonyl-l-isoleucine,l-isoleucine, n-phenylmethoxy carbonyl,2s,3s-2-benzyloxy carbonyl amino-3-methylpentanoic acid PubChem CID: 2724772 IUPAC Name: (2S,3S)-3-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid SMILES: CCC(C)C(C(=O)O)NC(=O)OCC1=CC=CC=C1