missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-glutamine 98.0+%, TCI America™

Catalog No. p-7124505
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05745G 5 g
C057425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05745G Supplier TCI America Supplier No. C05745G

Chemical Identifiers

CAS 2650-64-8
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00008043
InChI Key JIMLDJNLXLMGLX-JTQLQIEISA-N
Synonym z-gln-oh, n-carbobenzyloxy-l-glutamine, cbz-l-glutamine, cbz-gln-oh, n-carbobenzoxy-l-glutamine, benzyloxycarbonyl-l-glutamine, carbobenzoxyglutamine, n-cbz-l-glutamine, cbz-l-gln-oh, carbobenzoxy-l-glutamine
PubChem CID 75855
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-4-carbamoylbutanoic acid
SMILES NC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 138°C
Color White
Quantity 5 g
Formula Weight 280.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-glutamine