accessibility menu, dialog, popup

UserName

H2NO(S)NHOOOHO

CAS RN 2650-64-8

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-4-carbamoylbutanoic acid
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}-4-carbamoylbutanoic acid
Molecular Weight (g/mol): 280.28
Molecular Formula: C13H16N2O5
InChi Key: JIMLDJNLXLMGLX-JTQLQIEISA-N
SMILES: NC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)