missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-2-phenylglycine 98.0+%, TCI America™

Catalog No. C25671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25671G 1 g
C25675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25671G Supplier TCI America Supplier No. C25671G

Chemical Identifiers

CAS 53990-33-3
Molecular Formula C16H15NO4
Molecular Weight (g/mol) 285.299
MDL Number MFCD00077033
InChI Key RLDJWBVOZVJJOS-AWEZNQCLSA-N
Synonym z-phg-oh, z-l-phenylglycine, s-2-benzyloxycarbonylamino-2-phenylacetic acid, cbz-phg-oh, cbz-l-+-phenylglycine, z-l-2-phenylglycine, s-2-benzyloxy carbonyl amino-2-phenylacetic acid, z-l-phg-oh, 2s-benzyloxy carbonyl amino phenyl acetic acid, n-carbobenzoxy-l-2-phenylglycine
PubChem CID 819214
IUPAC Name (2S)-2-phenyl-2-(phenylmethoxycarbonylamino)acetic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 133°C
Color White
Quantity 1 g
Formula Weight 285.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-2-phenylglycine