missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-DL-phenylalanine 98.0+%, TCI America™
$69.31 - $138.61
Chemical Identifiers
| CAS | 3588-57-6 |
|---|---|
| Molecular Formula | C17H17NO4 |
| Molecular Weight (g/mol) | 299.326 |
| MDL Number | MFCD00063150 |
| InChI Key | RRONHWAVOYADJL-UHFFFAOYSA-N |
| Synonym | N-Cbz-DL-phenylalanine, Z-DL-Phe-OH |
| PubChem CID | 100081 |
| IUPAC Name | 3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid |
| SMILES | C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2 |
Chemical Identifiers
| 3588-57-6 | |
| 299.326 | |
| RRONHWAVOYADJL-UHFFFAOYSA-N | |
| 100081 | |
| C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2 |
| C17H17NO4 | |
| MFCD00063150 | |
| N-Cbz-DL-phenylalanine, Z-DL-Phe-OH | |
| 3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid |
Specifications
| 3588-57-6 | |
| White | |
| MFCD00063150 | |
| N-Cbz-DL-phenylalanine, Z-DL-Phe-OH | |
| C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2 | |
| 299.326 | |
| 299.33 | |
| Crystalline Powder |
| 104°C | |
| C17H17NO4 | |
| 10 g | |
| RRONHWAVOYADJL-UHFFFAOYSA-N | |
| 3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid | |
| 100081 | |
| ≥98.0% (HPLC,T) | |
| N-Carbobenzoxy-DL-phenylalanine |
Safety and Handling
TSCA : No