CAS RN 3588-57-6
IUPAC Name:
2-{[(benzyloxy)carbonyl]amino}-3-phenylpropanoic acid
Synonyms:
carbobenzoxyphenylalanine
Molecular Weight (g/mol):
299.33
Molecular Formula:
C17H17NO4
InChi Key:
RRONHWAVOYADJL-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N-Carbobenzoxy-DL-phenylalanine 98.0+%, TCI America™
CAS: 3588-57-6 Molecular Formula: C17H17NO4 Molecular Weight (g/mol): 299.326 MDL Number: MFCD00063150 InChI Key: RRONHWAVOYADJL-UHFFFAOYSA-N Synonym: N-Cbz-DL-phenylalanine, Z-DL-Phe-OH PubChem CID: 100081 IUPAC Name: 3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid SMILES: C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2