missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-DL-phenylalanine 98.0+%, TCI America™

Catalog No. C064325G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C064325G 25 g
C064310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C064325G Supplier TCI America Supplier No. C064325G

Chemical Identifiers

CAS 3588-57-6
Molecular Formula C17H17NO4
Molecular Weight (g/mol) 299.326
MDL Number MFCD00063150
InChI Key RRONHWAVOYADJL-UHFFFAOYSA-N
Synonym N-Cbz-DL-phenylalanine, Z-DL-Phe-OH
PubChem CID 100081
IUPAC Name 3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 104°C
Color White
Quantity 25 g
Formula Weight 299.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-phenylalanine