missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-DL-phenylalanine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 104°C |
| Color | White |
| Quantity | 10 g |
| Formula Weight | 299.33 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Carbobenzoxy-DL-phenylalanine |
Safety and Handling
| TSCA | No |