missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-D-valine 98.0+%, TCI America™

Catalog No. p-7125307
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21395G 5 g
C213925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21395G Supplier TCI America Supplier No. C21395G
Only null left

Chemical Identifiers

CAS 1685-33-2
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00065703
InChI Key CANZBRDGRHNSGZ-LLVKDONJSA-N
Synonym z-d-val-oh, cbz-d-valine, n-benzyloxycarbonyl-d-valine, cbz-d-val-oh, n-cbz-d-valine, z-d-valine, n-carbobenzoxy-d-valine, benzyloxycarbonyl-d-valine, r-2-benzyloxy carbonyl amino-3-methylbutanoic acid, d-valine, n-phenylmethoxy carbonyl
PubChem CID 712434
IUPAC Name (2R)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
SMILES CC(C)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 61°C
Color White
Quantity 5 g
Formula Weight 251.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-valine