accessibility menu, dialog, popup

UserName

H3CCH3(R)OOHNHOO

CAS RN 1685-33-2

IUPAC Name: (2R)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
Synonyms: (2R)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid|Z-D-VAL-OH
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: CANZBRDGRHNSGZ-LLVKDONJSA-N
SMILES: CC(C)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)