missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-D-proline 98.0+%, TCI America™
$78.29 - $225.56
Chemical Identifiers
| CAS | 6404-31-5 |
|---|---|
| Molecular Formula | C13H15NO4 |
| Molecular Weight (g/mol) | 249.266 |
| MDL Number | MFCD00063228 |
| InChI Key | JXGVXCZADZNAMJ-LLVKDONJSA-N |
| Synonym | n-benzyloxycarbonyl-d-proline, z-d-pro-oh, +-z-d-proline, z-d-proline, r-n-benzyloxycarbonylproline, n-cbz-d-proline, n-carbobenzoxy-d-proline, 1-benzyloxy carbonyl-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid |
| PubChem CID | 111208 |
| IUPAC Name | (2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid |
| SMILES | C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
Chemical Identifiers
| 6404-31-5 | |
| 249.266 | |
| JXGVXCZADZNAMJ-LLVKDONJSA-N | |
| 111208 | |
| C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
| C13H15NO4 | |
| MFCD00063228 | |
| n-benzyloxycarbonyl-d-proline, z-d-pro-oh, +-z-d-proline, z-d-proline, r-n-benzyloxycarbonylproline, n-cbz-d-proline, n-carbobenzoxy-d-proline, 1-benzyloxy carbonyl-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid | |
| (2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid |
Specifications
| 6404-31-5 | |
| C13H15NO4 | |
| 5 g | |
| JXGVXCZADZNAMJ-LLVKDONJSA-N | |
| (2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid | |
| 111208 | |
| ≥98.0% (HPLC,T) | |
| N-Carbobenzoxy-D-proline |
| 76°C | |
| MFCD00063228 | |
| n-benzyloxycarbonyl-d-proline, z-d-pro-oh, +-z-d-proline, z-d-proline, r-n-benzyloxycarbonylproline, n-cbz-d-proline, n-carbobenzoxy-d-proline, 1-benzyloxy carbonyl-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid | |
| C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O | |
| 249.266 | |
| 249.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6709
TSCA : No