accessibility menu, dialog, popup

UserName

CAS RN 6404-31-5

OO(R)NOO

CAS RN 6404-31-5

IUPAC Name: (2R)-1-[(benzyloxy)carbonyl]pyrrolidine-2-carboxylate
Synonyms: (2R)-1-[(benzyloxy)carbonyl]pyrrolidine-2-carboxylate
Molecular Weight (g/mol): 248.26
Molecular Formula: C13H14NO4
InChi Key: JXGVXCZADZNAMJ-LLVKDONJSA-M
SMILES: [O-]C(=O)[C@H]1CCCN1C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results