missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-D-methionine 98.0+%, TCI America™
$32.72 - $180.97
Chemical Identifiers
| CAS | 28862-80-8 |
|---|---|
| Molecular Formula | C13H17NO4S |
| Molecular Weight (g/mol) | 283.34 |
| MDL Number | MFCD00026043 |
| InChI Key | FPKHNNQXKZMOJJ-LLVKDONJSA-N |
| Synonym | z-d-met-oh, n-cbz-d-methionine, cbz-d-methionine, z-d-methionine, n-carbobenzoxy-d-methionine, r-2-benzyloxy carbonyl amino-4-methylthio butanoic acid, z-l-methionine, carbobenzyloxy-d-methionine, 2r-2-benzyloxy carbonyl amino-4-methylsulfanyl butanoic acid, pubchem14967 |
| PubChem CID | 1712153 |
| IUPAC Name | (2R)-2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid |
| SMILES | CSCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C0665100MG
|
TCI America
C0665100MG |
100 mg |
Each for $32.72
|
|
|||||
|
C06655G
|
TCI America
C06655G |
5 g |
Each for $59.69
|
|
|||||
|
C066525G
|
TCI America
C066525G |
25 g |
Each for $180.97
|
|
|||||
Chemical Identifiers
| 28862-80-8 | |
| 283.34 | |
| FPKHNNQXKZMOJJ-LLVKDONJSA-N | |
| 1712153 | |
| CSCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
| C13H17NO4S | |
| MFCD00026043 | |
| z-d-met-oh, n-cbz-d-methionine, cbz-d-methionine, z-d-methionine, n-carbobenzoxy-d-methionine, r-2-benzyloxy carbonyl amino-4-methylthio butanoic acid, z-l-methionine, carbobenzyloxy-d-methionine, 2r-2-benzyloxy carbonyl amino-4-methylsulfanyl butanoic acid, pubchem14967 | |
| (2R)-2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid |
Specifications
| 28862-80-8 | |
| White | |
| MFCD00026043 | |
| z-d-met-oh, n-cbz-d-methionine, cbz-d-methionine, z-d-methionine, n-carbobenzoxy-d-methionine, r-2-benzyloxy carbonyl amino-4-methylthio butanoic acid, z-l-methionine, carbobenzyloxy-d-methionine, 2r-2-benzyloxy carbonyl amino-4-methylsulfanyl butanoic acid, pubchem14967 | |
| CSCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O | |
| 283.34 | |
| 283.34 | |
| Crystalline Powder |
| 69°C | |
| C13H17NO4S | |
| 100 mg | |
| FPKHNNQXKZMOJJ-LLVKDONJSA-N | |
| (2R)-2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid | |
| 1712153 | |
| ≥98.0% (T) | |
| N-Carbobenzoxy-D-methionine |
Safety and Handling
TSCA : No