accessibility menu, dialog, popup

UserName

H3CS(R)OHONHOO

CAS RN 28862-80-8

IUPAC Name: (2R)-2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Synonyms: (2R)-2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid|Z-D-MET-OH
Molecular Weight (g/mol): 283.34
Molecular Formula: C13H17NO4S
InChi Key: FPKHNNQXKZMOJJ-LLVKDONJSA-N
SMILES: CSCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)