missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-D-methionine 98.0+%, TCI America™

Catalog No. C0665100MG
Change view
Click to view available options
Quantity:
100 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0665100MG 100 mg
C06655G 5 g
C066525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0665100MG Supplier TCI America Supplier No. C0665100MG

Chemical Identifiers

CAS 28862-80-8
Molecular Formula C13H17NO4S
Molecular Weight (g/mol) 283.34
MDL Number MFCD00026043
InChI Key FPKHNNQXKZMOJJ-LLVKDONJSA-N
Synonym z-d-met-oh, n-cbz-d-methionine, cbz-d-methionine, z-d-methionine, n-carbobenzoxy-d-methionine, r-2-benzyloxy carbonyl amino-4-methylthio butanoic acid, z-l-methionine, carbobenzyloxy-d-methionine, 2r-2-benzyloxy carbonyl amino-4-methylsulfanyl butanoic acid, pubchem14967
PubChem CID 1712153
IUPAC Name (2R)-2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 69°C
Color White
Quantity 100 mg
Formula Weight 283.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-methionine