missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-beta-alanine 98.0+%, TCI America™
$32.28 - $108.21
Chemical Identifiers
| CAS | 2304-94-1 |
|---|---|
| Molecular Formula | C11H13NO4 |
| Molecular Weight (g/mol) | 223.23 |
| MDL Number | MFCD00037292 |
| InChI Key | GEVGRLPYQJTKKS-UHFFFAOYSA-N |
| Synonym | z-beta-ala-oh, carbobenzyloxy-beta-alanine, n-cbz-beta-alanine, cbz-beta-alanine, n-benzyloxycarbonyl-beta-alanine, n-cbz-b-alanine, 3-benzyloxy carbonyl amino propanoic acid, cbz-beta-ala-oh, n-carbobenzoxy-beta-alanine |
| PubChem CID | 75313 |
| IUPAC Name | 3-{[(benzyloxy)carbonyl]amino}propanoic acid |
| SMILES | OC(=O)CCNC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 2304-94-1 | |
| 223.23 | |
| GEVGRLPYQJTKKS-UHFFFAOYSA-N | |
| 75313 | |
| OC(=O)CCNC(=O)OCC1=CC=CC=C1 |
| C11H13NO4 | |
| MFCD00037292 | |
| z-beta-ala-oh, carbobenzyloxy-beta-alanine, n-cbz-beta-alanine, cbz-beta-alanine, n-benzyloxycarbonyl-beta-alanine, n-cbz-b-alanine, 3-benzyloxy carbonyl amino propanoic acid, cbz-beta-ala-oh, n-carbobenzoxy-beta-alanine | |
| 3-{[(benzyloxy)carbonyl]amino}propanoic acid |
Specifications
| 105°C | |
| White | |
| MFCD00037292 | |
| GEVGRLPYQJTKKS-UHFFFAOYSA-N | |
| 3-{[(benzyloxy)carbonyl]amino}propanoic acid | |
| 223.23 | |
| 223.23 | |
| Crystalline Powder |
| 2304-94-1 | |
| C11H13NO4 | |
| z-beta-ala-oh, carbobenzyloxy-beta-alanine, n-cbz-beta-alanine, cbz-beta-alanine, n-benzyloxycarbonyl-beta-alanine, n-cbz-b-alanine, 3-benzyloxy carbonyl amino propanoic acid, cbz-beta-ala-oh, n-carbobenzoxy-beta-alanine | |
| OC(=O)CCNC(=O)OCC1=CC=CC=C1 | |
| 1 g | |
| 75313 | |
| ≥98.0% (T) | |
| N-Carbobenzoxy-beta-alanine |
Safety and Handling
TSCA : No