missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-beta-alanine 98.0+%, TCI America™
Supplier: TCI America C064010G
Specifications
| 105°C | |
| 2304-94-1 | |
| C11H13NO4 | |
| z-beta-ala-oh, carbobenzyloxy-beta-alanine, n-cbz-beta-alanine, cbz-beta-alanine, n-benzyloxycarbonyl-beta-alanine, n-cbz-b-alanine, 3-benzyloxy carbonyl amino propanoic acid, cbz-beta-ala-oh, n-carbobenzoxy-beta-alanine | |
| OC(=O)CCNC(=O)OCC1=CC=CC=C1 | |
| 10 g | |
| 75313 | |
| ≥98.0% (T) |
| N-Carbobenzoxy-beta-alanine | |
| White | |
| MFCD00037292 | |
| GEVGRLPYQJTKKS-UHFFFAOYSA-N | |
| 3-{[(benzyloxy)carbonyl]amino}propanoic acid | |
| 223.23 | |
| 223.23 | |
| Crystalline Powder |
Chemical Identifiers
| 2304-94-1 | |
| 223.23 | |
| GEVGRLPYQJTKKS-UHFFFAOYSA-N | |
| 75313 | |
| OC(=O)CCNC(=O)OCC1=CC=CC=C1 |
| C11H13NO4 | |
| MFCD00037292 | |
| z-beta-ala-oh, carbobenzyloxy-beta-alanine, n-cbz-beta-alanine, cbz-beta-alanine, n-benzyloxycarbonyl-beta-alanine, n-cbz-b-alanine, 3-benzyloxy carbonyl amino propanoic acid, cbz-beta-ala-oh, n-carbobenzoxy-beta-alanine | |
| 3-{[(benzyloxy)carbonyl]amino}propanoic acid |
Safety and Handling
TSCA : No