missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-1,5-diaminopentane Hydrochloride 98.0+%, TCI America™

Catalog No. C15201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15201G 1 g
C15205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15201G Supplier TCI America Supplier No. C15201G

Chemical Identifiers

CAS 18807-74-4
Molecular Formula C13H21ClN2O2
Molecular Weight (g/mol) 272.773
MDL Number MFCD00270152
InChI Key VYIRBXGDTOPWSY-UHFFFAOYSA-N
Synonym N-(5-Aminoamyl)carbamic Acid Benzyl Ester Hydrochloride, N-(5-Aminopentyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl N-(5-Aminoamyl)carbamate Hydrochloride, Benzyl N-(5-Aminopentyl)carbamate Hydrochloride, N-Cbz-1,5-diaminopentane Hydrochloride
PubChem CID 13196228
IUPAC Name benzyl N-(5-aminopentyl)carbamate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)NCCCCCN.Cl

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 1 g
Formula Weight 272.77
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-1,5-diaminopentane Hydrochloride