accessibility menu, dialog, popup

UserName

OOHNNH2HCl

CAS RN 18807-74-4

IUPAC Name: benzyl N-(5-aminopentyl)carbamate hydrochloride
Synonyms: benzyl N-(5-aminopentyl)carbamate hydrochloride
Molecular Weight (g/mol): 272.77
Molecular Formula: C13H21ClN2O2
InChi Key: VYIRBXGDTOPWSY-UHFFFAOYSA-N
SMILES: Cl.NCCCCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)